Thermo Scientific Chemicals

2-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 344660010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 344660010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 344660010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5720-06-9
IUPAC Name
(2-methoxyphenyl)boronic acid
Molecular Formula
C7H9BO3
InChI Key
ROEQGIFOWRQYHD-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1B(O)O
Appearance (Color)
Off-white
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Titration with NaOH
>=96.0 %
Melting point
106°C to 111°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text