Thermo Scientific Chemicals

Methylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 344670050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 344670050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 344670050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13061-96-6
IUPAC Name
methylboronic acid
Molecular Formula
CH5BO2
InChI Key
KTMKRRPZPWUYKK-UHFFFAOYSA-N
SMILES
CB(O)O
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
87°C to 91°C
Titration with NaOH
96.0 to 104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text