1-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals
1-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 344690050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13922-41-3
IUPAC Name(naphthalen-1-yl)boronic acid
Molecular FormulaC10H9BO2
InChI KeyHUMMCEUVDBVXTQ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to pink
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only