Thermo Scientific Chemicals

4-Aminopyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 345070010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Aminopyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 345070010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 345070010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
591-54-8
IUPAC Name
pyrimidin-4-amine
Molecular Formula
C4H5N3
InChI Key
OYRRZWATULMEPF-UHFFFAOYSA-N
SMILES
NC1=CC=NC=N1
Appearance (Color)
White to light beige
Appearance (Form)
Powder or crystals
HPLC
>=97.5 %
Infrared spectrum
Conforms
Melting point
151°C to 155°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text