Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Sterility | Capacity (Metric) | Unit Size |
---|---|---|---|
3451 | Sterile | 1.5 mL | Case of 5000 |
3453 | Sterile | 2 mL | Case of 5000 |
3453PK | Sterile | 2 mL | Pack of 500 |
3434 | Non-sterile | 2 mL | Case of 5000 |
3434PK | Non-sterile | 2 mL | Pack of 500 |
3451PK | Sterile | 1.5 mL | Pack of 500 |
3446 | Non-sterile | 0.6 mL | Case of 10000 |
3446PK | Non-sterile | 0.6 mL | Pack of 1000 |
3449 | Sterile | 0.6 mL | Case of 10000 |
3449PK also known as 3449 | Sterile | 0.6 mL | Pack of 1000 |
Improve sample recovery by significantly reducing surface binding with Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes. Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. With no additives or coatings, these tubes — available in 0.6mL, 1.5mL and 2.0mL sizes — offer maximum sample recovery by significantly reducing surface binding.
Color/Packaging Options
Highest Quality
Recommended for:
Daily laboratory usage including centrifugation and short-term sample storage
Share catalog number, name or link