N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%, Thermo Scientific Chemicals
N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 345180100
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.50 
Save 11.85 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.50 
Save 11.85 (15%)
Add to cart
N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%, Thermo Scientific Chemicals
Catalog number345180100
Price (USD)/ Each
68.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
80.50 
Save 11.85 (15%)
-
Add to cart
Chemical Identifiers
CAS4439-20-7
IUPAC Name2-({2-[(2-hydroxyethyl)amino]ethyl}amino)ethan-1-ol
Molecular FormulaC6H16N2O2
InChI KeyGFIWSSUBVYLTRF-UHFFFAOYSA-N
SMILESOCCNCCNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point94°C to 101°C
Appearance (Form)Powder or crystals and/or chunks
Appearance (Color)White to light yellow to beige
Infrared spectrumConforms
Titration with HClO494 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only