Thermo Scientific Chemicals

N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%, Thermo Scientific Chemicals

Catalog number: 345180500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%, Thermo Scientific Chemicals

Catalog number: 345180500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 345180500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4439-20-7
IUPAC Name
2-({2-[(2-hydroxyethyl)amino]ethyl}amino)ethan-1-ol
Molecular Formula
C6H16N2O2
InChI Key
GFIWSSUBVYLTRF-UHFFFAOYSA-N
SMILES
OCCNCCNCCO
Melting point
94°C to 101°C
Appearance (Form)
Powder or crystals and/or chunks
Appearance (Color)
White to light yellow to beige
Infrared spectrum
Conforms
Titration with HClO4
94 to 106 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text