N,N-Dimethylglycine, 97%
N,N-Dimethylglycine, 97%
Thermo Scientific Chemicals

N,N-Dimethylglycine, 97%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 345200050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1118-68-9
IUPAC Name2-(dimethylamino)acetic acid
Molecular FormulaC4H9NO2
InChI KeyFFDGPVCHZBVARC-UHFFFAOYSA-N
SMILESCN(C)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Melting point175°C to 181°C
Appearance (Form)Crystalline powder
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only