Thermo Scientific Chemicals

Iron(III) sulfate pentahydrate, 97%, Thermo Scientific Chemicals

Catalog number: 345235000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Iron(III) sulfate pentahydrate, 97%, Thermo Scientific Chemicals

Catalog number: 345235000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 345235000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142906-29-4
IUPAC Name
diiron(3+) pentahydrate trisulfate
Molecular Formula
Fe2H10O17S3
InChI Key
YHGPYBQVSJBGHH-UHFFFAOYSA-H
SMILES
O.O.O.O.O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Appearance (Form)
Crystalline or granular powder
Insoluble matter
=<0.01 % (in HCl)
Zinc (Zn)
=<50 ppm
Subst not pptd by NH3
=<0.05 % (as SO4)
Copper (Cu)
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text