Methylcyclopentane, 95%
Methylcyclopentane, 95%
Methylcyclopentane, 95%
Methylcyclopentane, 95%
Thermo Scientific Chemicals

Methylcyclopentane, 95%

CAS: 96-37-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 L
Packaging:
View all
Glass bottle
Catalog number 345245000
Price (USD)
141.65
Special offer
Online exclusive
Ends: 26-Sep-2025
175.00
Save 33.35 (19%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
141.65
Special offer
Online exclusive
Ends: 26-Sep-2025
175.00
Save 33.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS96-37-7
IUPAC Namemethylcyclopentane
Molecular FormulaC6H12
InChI KeyGDOPTJXRTPNYNR-UHFFFAOYSA-N
SMILESCC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=94 %
Appearance (Form)Liquid
Refractive index1.4070 to 1.4110 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link