3-Amino-5-nitrobenzoisothiazole, 95%, Thermo Scientific Chemicals
3-Amino-5-nitrobenzoisothiazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-5-nitrobenzoisothiazole, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog number 345410250
Price (USD)/ Each
-
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-nitro-1,2-benzothiazol-3-amine
Molecular FormulaC7H5N3O2S
InChI KeyLDTCWISGJYTXDC-UHFFFAOYSA-N
SMILESNC1=NSC2=CC=C(C=C12)[N+]([O-])=O
Molecular Weight (g/mol)195.20
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red
Infrared spectrumConforms
Titration Total base>=94 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only