Thermo Scientific Chemicals

2-Chloro-4-aminopyridine, 97%, Thermo Scientific Chemicals

Catalog number: 345450250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Chloro-4-aminopyridine, 97%, Thermo Scientific Chemicals

Catalog number: 345450250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 345450250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14432-12-3
IUPAC Name
2-chloropyridin-4-amine
Molecular Formula
C5H5ClN2
InChI Key
BLBDTBCGPHPIJK-UHFFFAOYSA-N
SMILES
NC1=CC=NC(Cl)=C1
Appearance (Form)
Crystalline powder
Titration with HClO4
>=96.0 %
Melting point
89°C to 94°C
Appearance (Color)
Light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text