Thermo Scientific Chemicals

1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals

Catalog number: 345480050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals

Catalog number: 345480050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 345480050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
288-36-8
IUPAC Name
2H-1,2,3-triazole
Molecular Formula
C2H3N3
InChI Key
QWENRTYMTSOGBR-UHFFFAOYSA-N
SMILES
N1N=CC=N1
Appearance (Form)
Liquid
Refractive index
1.4940 to 1.4980 (20°C, 589 nm)
Infrared spectrum
Conforms
Titration with HClO4
96.0 to 104.0 %
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text