3-Thiopheneboronic acid, 97%, Thermo Scientific Chemicals
3-Thiopheneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Thiopheneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 345660010
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.30 
Save 4.65 (14%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.30 
Save 4.65 (14%)
Add to cart
3-Thiopheneboronic acid, 97%, Thermo Scientific Chemicals
Catalog number345660010
Price (USD)/ Each
28.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
33.30 
Save 4.65 (14%)
-
Add to cart
Chemical Identifiers
CAS6165-69-1
IUPAC Name(thiophen-3-yl)boronic acid
Molecular FormulaC4H5BO2S
InChI KeyQNMBSXGYAQZCTN-UHFFFAOYSA-N
SMILESOB(O)C1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Titration with NaOH96.0 to 106.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only