Thermo Scientific Chemicals

3-Thiopheneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 345660050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Thiopheneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 345660050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 345660050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6165-69-1
IUPAC Name
(thiophen-3-yl)boronic acid
Molecular Formula
C4H5BO2S
InChI Key
QNMBSXGYAQZCTN-UHFFFAOYSA-N
SMILES
OB(O)C1=CSC=C1
Appearance (Color)
White to beige to grey
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Form)
Powder
Titration with NaOH
96.0 to 106.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text