Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 1.0M sol. in methyl tert-butyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 345671000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 1.0M sol. in methyl tert-butyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 345671000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 345671000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4039-32-1, 1634-04-4
Titration Active base
21 to 25 % (as LiNSi)
Appearance (Color)
Clear to hazy yellow to brown
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text