Thermo Scientific Chemicals

Trimethyl borate, 99%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 346128000
800 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Trimethyl borate, 99%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 346128000
800 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 346128000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
121-43-7
IUPAC Name
trimethyl borate
Molecular Formula
C3H9BO3
InChI Key
WRECIMRULFAWHA-UHFFFAOYSA-N
SMILES
COB(OC)OC
Titration with NaOH
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text