Thermo Scientific Chemicals

3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech., Thermo Scientific Chemicals

Catalog number: 346171000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech., Thermo Scientific Chemicals

Catalog number: 346171000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 346171000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
35141-30-1
IUPAC Name
3,3-dimethoxy-2-oxa-7,10-diaza-3-siladodecan-12-amine
Molecular Formula
C10H27N3O3Si
InChI Key
NHBRUUFBSBSTHM-UHFFFAOYSA-N
SMILES
CO[Si](CCCNCCNCCN)(OC)OC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4570 to 1.4650 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Appearance
May discolor to pink to red to brown on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text