all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized, Thermo Scientific Chemicals
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 g
Catalog number 346325000
Price (USD)/ Each
490.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
577.00 
Save 86.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
490.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
577.00 
Save 86.35 (15%)
Add to cart
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized, Thermo Scientific Chemicals
Catalog number346325000
Price (USD)/ Each
490.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
577.00 
Save 86.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate
Molecular FormulaC22H32O2
InChI KeyQGNJRVVDBSJHIZ-QHLGVNSISA-N
SMILESCC(=O)OCC=C(/C)C=CC=C(/C)C=CC1=C(C)CCCC1(C)C
Molecular Weight (g/mol)328.50
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Oily liquid
Assay1.50 to 1.65 million IU/g (Vitamin A)
Peroxide value=<15 meq/kg
Related substancesper USP
Related substances=<0.54 (A350/A326)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only