Thermo Scientific Chemicals

1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate, pure, indicator gra

Catalog number: 346551000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate, pure, indicator gra

Catalog number: 346551000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 346551000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
525-05-3
IUPAC Name
disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
Molecular Formula
C10H5NNa2O8S2
InChI Key
DMKMTGULLYISBH-UHFFFAOYSA-L
SMILES
[Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
Loss on drying
=<5 % (1 g, 105°C)
Appearance (Form)
Crystalline powder
Appearance (Color)
Yellow to orange
Infrared spectrum
Conforms
Suitability
(for Co determination) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text