1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate, pure, indicator gra
1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate, pure, indicator gra
Thermo Scientific Chemicals

1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate, pure, indicator gra

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 346555000
Price (USD)/ Each
266.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
314.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
266.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
314.00 
Save 47.35 (15%)
Add to cart
1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate, pure, indicator gra
Catalog number346555000
Price (USD)/ Each
266.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
314.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
CAS525-05-3
IUPAC Namedisodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
Molecular FormulaC10H5NNa2O8S2
InChI KeyDMKMTGULLYISBH-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<5 % (1 g, 105°C)
Appearance (Form)Crystalline powder
Appearance (Color)Yellow to orange
Infrared spectrumConforms
Suitability(for Co determination) Passes test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only