Thermo Scientific Chemicals

trans-4-Aminocyclohexanol, 97%, Thermo Scientific Chemicals

Catalog number: 346680050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

trans-4-Aminocyclohexanol, 97%, Thermo Scientific Chemicals

Catalog number: 346680050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 346680050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
27489-62-9
IUPAC Name
4-aminocyclohexan-1-ol
Molecular Formula
C6H13NO
InChI Key
IMLXLGZJLAOKJN-UHFFFAOYSA-N
SMILES
NC1CCC(O)CC1
Appearance (Form)
Powder or crystals
Melting point
108°C to 113°C
Infrared spectrum
Conforms
Appearance (Color)
White to cream to light brown
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text