Thermo Scientific Chemicals

N-Pivaloyl-o-toluidine, 99%, Thermo Scientific Chemicals

Catalog number: 346730050
5 g, Each
Thermo Scientific Chemicals

N-Pivaloyl-o-toluidine, 99%, Thermo Scientific Chemicals

Catalog number: 346730050
5 g, Each
Quantity
Catalog number: 346730050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
61495-04-3
IUPAC Name
2,2-dimethyl-N-(2-methylphenyl)propanamide
Molecular Formula
C12H17NO
InChI Key
CSGRQLUGMVFNON-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1NC(=O)C(C)(C)C
Appearance (Color)
White
Melting point
111°C to 115°C
Appearance (Form)
Crystalline powder
GC
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text