Thermo Scientific Chemicals

4-Bromobenzyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 346750250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromobenzyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 346750250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 346750250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
589-17-3
IUPAC Name
1-bromo-4-(chloromethyl)benzene
Molecular Formula
C7H6BrCl
InChI Key
BSIIGUGKOPPTPZ-UHFFFAOYSA-N
SMILES
ClCC1=CC=C(Br)C=C1
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
36°C to 40°C
Appearance (Color)
White to light beige
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text