Thermo Scientific Chemicals

Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 346760250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 346760250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 346760250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
531-81-7
IUPAC Name
2-oxo-2H-chromene-3-carboxylic acid
Molecular Formula
C10H6O4
InChI Key
ACMLKANOGIVEPB-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC2=CC=CC=C2OC1=O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Appearance (Color)
Light brown
Titration with TBAH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text