Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 346761000
Price (USD)/ Each
184.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
184.00
Add to cart
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Catalog number346761000
Price (USD)/ Each
184.00
-
Add to cart
Chemical Identifiers
CAS531-81-7
IUPAC Name2-oxo-2H-chromene-3-carboxylic acid
Molecular FormulaC10H6O4
InChI KeyACMLKANOGIVEPB-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=CC=CC=C2OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Light brown
Titration with TBAH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only