Thermo Scientific Chemicals

Acriflavine hydrochloride, Thermo Scientific Chemicals

Catalog number: 346841000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Acriflavine hydrochloride, Thermo Scientific Chemicals

Catalog number: 346841000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 346841000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69235-50-3
IUPAC Name
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride
Molecular Formula
C27H27Cl3N6
InChI Key
MKLTXAHQKDVBLY-UHFFFAOYSA-N
SMILES
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
Appearance (Form)
Powder
Loss on drying
=<7 % (105°C, 1 h)
Appearance (Color)
Orange-red to red-brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text