Thermo Scientific Chemicals

Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3, Thermo Scientific Chemicals

Catalog number: 346930050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3, Thermo Scientific Chemicals

Catalog number: 346930050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 346930050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15110-17-5
IUPAC Name
diethyl (3,3-diethoxypropyl)phosphonate
Molecular Formula
C11H25O5P
InChI Key
WKPXMGJOGYEGNW-UHFFFAOYSA-N
SMILES
CCOC(CCP(=O)(OCC)OCC)OCC
Refractive index
1.4290 to 1.4320 (20°C, 589 nm)
GC
>=94 %
Appearance (Color)
Clear light yellow to orange
Stabilizer
=<1 % Na2CO3
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text