Thermo Scientific Chemicals

Methyltrioxorhenium (VII), 98%, Thermo Scientific Chemicals

Catalog number: 346960025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyltrioxorhenium (VII), 98%, Thermo Scientific Chemicals

Catalog number: 346960025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 346960025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70197-13-6
IUPAC Name
trioxorhenium methanide
Molecular Formula
CH3O3Re
InChI Key
VZSXFJPZOCRDPW-UHFFFAOYSA-N
SMILES
[CH3-].O=[Re](=O)=O
Appearance (Form)
Powder or crystals or needles and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text