Thermo Scientific Chemicals

D-Threonine, 98%

Catalog number: 346970250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

D-Threonine, 98%

Catalog number: 346970250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 346970250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
632-20-2
IUPAC Name
(2R,3S)-2-amino-3-hydroxybutanoic acid
Molecular Formula
C4H9NO3
InChI Key
AYFVYJQAPQTCCC-STHAYSLISA-N
SMILES
C[C@H](O)[C@@H](N)C(O)=O
Appearance (Form)
Crystalline powder or crystals
Heavy metals (as Pb)
=<10 ppm
Chloride (Cl)
=<200 ppm
Sulfate (SO4)
=<300 ppm
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text