Thermo Scientific Chemicals

2-Bromo-5-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: 347170250
25 g, Each
Thermo Scientific Chemicals

2-Bromo-5-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: 347170250
25 g, Each
Quantity
Catalog number: 347170250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3510-66-5
IUPAC Name
2-bromo-5-methylpyridine
Molecular Formula
C6H6BrN
InChI Key
YWNJQQNBJQUKME-UHFFFAOYSA-N
SMILES
CC1=CC=C(Br)N=C1
Infrared spectrum
Conforms
Melting point
40°C to 45°C
Appearance (Color)
White to light yellow to light brown
Appearance (Form)
Low melting crystalline solid or crystals
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text