Thermo Scientific Chemicals

Potassium carbonate sesquihydrate, 98.5%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 347200025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium carbonate sesquihydrate, 98.5%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 347200025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 347200025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6381-79-9
IUPAC Name
tetrapotassium trihydrate dicarbonate
Molecular Formula
C2H6K4O9
InChI Key
WWVGGBSGYNQKCY-UHFFFAOYSA-J
SMILES
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
Insoluble matter
=<0.01 %
Phosphate (PO4)
=<0.001 %
Heavy metals (ICP-OES)
=<5 ppm
Titration with HCl
98.5 to 101.0 %
Sulfur compounds (as SO4)
=<0.004 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text