Thermo Scientific Chemicals

(S)-cis-Verbenol, 97%, sum of isomers, Thermo Scientific Chemicals

Catalog number: 347330050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(S)-cis-Verbenol, 97%, sum of isomers, Thermo Scientific Chemicals

Catalog number: 347330050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 347330050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18881-04-4
IUPAC Name
(1S,2R,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol
Molecular Formula
C10H16O
InChI Key
WONIGEXYPVIKFS-DJLDLDEBSA-N
SMILES
CC1=C[C@@H](O)[C@H]2C[C@@H]1C2(C)C
GC
>=96.0 % (sum of isomers)
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Crystals
Specific optical rotation
+9° to +11° (20°C, 589 nm) (C=2, ethyl alcohol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text