Thermo Scientific Chemicals

4-Bromo-2-fluorobenzoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 347460250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 347460250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 347460250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
151982-51-3
IUPAC Name
4-bromo-2-fluorobenzoyl chloride
Molecular Formula
C7H3BrClFO
InChI Key
PCFIABOQFAFDAU-UHFFFAOYSA-N
SMILES
FC1=CC(Br)=CC=C1C(Cl)=O
Appearance (Form)
Crystalline mass or liquid
GC
>=98.5 %
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text