8-Hydroxyquinoline-2-carbonitrile, 98%, Thermo Scientific Chemicals
8-Hydroxyquinoline-2-carbonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Hydroxyquinoline-2-carbonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 347640010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6759-78-0
IUPAC Name8-hydroxyquinoline-2-carbonitrile
Molecular FormulaC10H6N2O
InChI KeyKUQKKIBQVSFDHX-UHFFFAOYSA-N
SMILESOC1=C2N=C(C=CC2=CC=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Melting point131°C to 136°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only