Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number 347715000
Price (USD)/ Each
74.65
Online exclusive
82.30 
Save 7.65 (9%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
74.65
Online exclusive
82.30 
Save 7.65 (9%)
Add to cart
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Catalog number347715000
Price (USD)/ Each
74.65
Online exclusive
82.30 
Save 7.65 (9%)
-
Add to cart
Chemical Identifiers
CAS142-90-5
IUPAC Namedodecyl 2-methylprop-2-enoate
Molecular FormulaC16H30O2
InChI KeyGMSCBRSQMRDRCD-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCOC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4440 to 1.4470 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only