Thermo Scientific Chemicals

Dimethyltin dichloride, 95%, Thermo Scientific Chemicals

Catalog number: 347820050
5 g, Each
Thermo Scientific Chemicals

Dimethyltin dichloride, 95%, Thermo Scientific Chemicals

Catalog number: 347820050
5 g, Each
Quantity
Catalog number: 347820050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
753-73-1
Molecular Formula
C2H8Cl2Sn
InChI Key
QLQTYMCOTSXVHH-UHFFFAOYSA-N
SMILES
Cl.Cl.C[SnH2]C
Molecular Weight (g/mol)
221.70
Appearance (Color)
White to off-white
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Titration Argentometric
>=94 %
Melting point
103°C to 110°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text