Dimethyltin dichloride, 95%, Thermo Scientific Chemicals
Dimethyltin dichloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyltin dichloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 347820250
Price (USD)/ Each
132.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
156.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
132.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
156.00 
Save 23.35 (15%)
Add to cart
Dimethyltin dichloride, 95%, Thermo Scientific Chemicals
Catalog number347820250
Price (USD)/ Each
132.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
156.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
CAS753-73-1
Molecular FormulaC2H8Cl2Sn
InChI KeyQLQTYMCOTSXVHH-UHFFFAOYSA-N
SMILESCl.Cl.C[SnH2]C
Molecular Weight (g/mol)221.70
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration Argentometric>=94 %
Melting point103°C to 110°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only