Ferrocenemethanol, 97%, Thermo Scientific Chemicals
Ferrocenemethanol, 97%, Thermo Scientific Chemicals
Ferrocenemethanol, 97%, Thermo Scientific Chemicals
Ferrocenemethanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ferrocenemethanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 348030010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1273-86-5
Molecular FormulaC11H12FeO
SMILES[Fe].c1cccc1.OCc1cccc1
Molecular Weight (g/mol)216.06
MDL NumberMFCD00040878
SpecificationsSpecification SheetSpecification Sheet
Melting point77°C to 82°C
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)Yellow to golden to orange
Appearance (Form)Powder or needles
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection