Thermo Scientific Chemicals

1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 85%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 348070250
25 g, Each
Thermo Scientific Chemicals

1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 85%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 348070250
25 g, Each
Quantity
Catalog number: 348070250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4249-10-9
IUPAC Name
1,2,3,4-tetramethylcyclopenta-1,3-diene
Molecular Formula
C9H14
InChI Key
VNPQQEYMXYCAEZ-UHFFFAOYSA-N
SMILES
CC1=C(C)C(C)=C(C)C1
Appearance (Color)
Colorless to yellow to orange
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=80 % (mixture of isomers)
Refractive index
1.4700 to 1.4760 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text