Isobutylene oxide, 98%
Isobutylene oxide, 98%
Isobutylene oxide, 98%
Isobutylene oxide, 98%
Thermo Scientific Chemicals

Isobutylene oxide, 98%

CAS: 558-30-5 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 348501000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS558-30-5
IUPAC Name2,2-dimethyloxirane
Molecular FormulaC4H8O
InChI KeyGELKGHVAFRCJNA-UHFFFAOYSA-N
SMILESCC1(C)CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.3705 to 1.3745 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link