Toluidine Blue O, pure
Toluidine Blue O, pure
Thermo Scientific Chemicals

Toluidine Blue O, pure

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 348601000
Price (USD)/ Each
319.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
376.00 
Save 56.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
319.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
376.00 
Save 56.35 (15%)
Add to cart
Toluidine Blue O, pure
Catalog number348601000
Price (USD)/ Each
319.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
376.00 
Save 56.35 (15%)
-
Add to cart
Chemical Identifiers
CAS92-31-9
IUPAC Name3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
Molecular FormulaC15H16ClN3S
InChI KeyHNONEKILPDHFOL-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to black
Appearance (Form)Powder
Infrared spectrumConforms
UV spectrumComplies
NMRComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only