Methanesulfonic anhydride, 97%, Thermo Scientific Chemicals
Methanesulfonic anhydride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanesulfonic anhydride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 348760250
Price (USD)/ Each
69.65
Online exclusive
82.30 
Save 12.65 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
69.65
Online exclusive
82.30 
Save 12.65 (15%)
Add to cart
Methanesulfonic anhydride, 97%, Thermo Scientific Chemicals
Catalog number348760250
Price (USD)/ Each
69.65
Online exclusive
82.30 
Save 12.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethanesulfonyl methanesulfonate
Molecular FormulaC2H6O5S2
InChI KeyIZDROVVXIHRYMH-UHFFFAOYSA-N
SMILESCS(=O)(=O)OS(C)(=O)=O
Molecular Weight (g/mol)174.19
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration by coupling>=96.0 %
Appearance (Form)Powder or crystals and/or chunks
Appearance (Color)White to grey to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only