4-Hydroxypyridine, 95%, Thermo Scientific Chemicals
4-Hydroxypyridine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxypyridine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 348830250
Price (USD)/ Each
45.20
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.20
Add to cart
4-Hydroxypyridine, 95%, Thermo Scientific Chemicals
Catalog number348830250
Price (USD)/ Each
45.20
-
Add to cart
Chemical Identifiers
IUPAC Name1,4-dihydropyridin-4-one
Molecular FormulaC5H5NO
InChI KeyGCNTZFIIOFTKIY-UHFFFAOYSA-N
SMILESO=C1C=CNC=C1
Molecular Weight (g/mol)95.10
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=94 %
HPLC>=94 %
Melting point139°C to 151°C
Appearance (Color)White to yellow to beige to light brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only