1,2-O-Isopropylidene-alpha-D-xylofuranose, 99%
1,2-O-Isopropylidene-alpha-D-xylofuranose, 99%
Thermo Scientific Chemicals

1,2-O-Isopropylidene-alpha-D-xylofuranose, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 348880100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20031-21-4
IUPAC Name(3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Molecular FormulaC8H14O5
InChI KeyJAUQZVBVVJJRKM-XZBKPIIZSA-N
SMILESCC1(C)O[C@H]2O[C@H](CO)[C@H](O)[C@H]2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Specific optical rotation-17° to -19° (20°C, 589 nm) (c=1, water)
Appearance (Color)White
Appearance (Form)Waxy solid or coarse crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only