Thermo Scientific Chemicals

1,2-Dibromobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 348930250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Dibromobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 348930250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 348930250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
583-53-9
IUPAC Name
1,2-dibromobenzene
Molecular Formula
C6H4Br2
InChI Key
WQONPSCCEXUXTQ-UHFFFAOYSA-N
SMILES
BrC1=CC=CC=C1Br
Appearance (Form)
Liquid
Refractive index
1.6100 to 1.6120 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to light yellow to light pink
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text