Thermo Scientific Chemicals

4-(4,6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride, 97%, Thermo Scientific Chemicals

Catalog number: 348960050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(4,6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride, 97%, Thermo Scientific Chemicals

Catalog number: 348960050
5 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 348960050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3945-69-5
IUPAC Name
4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium chloride
Molecular Formula
C10H17ClN4O3
InChI Key
BMTZEAOGFDXDAD-UHFFFAOYSA-M
SMILES
[Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Appearance (Form)
Powder and/or chunks
Water
=<17 % (K.F.)
Titration with HClO4
>=96.0 % (on dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text