1,2,4-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
1,2,4-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,4-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
250 g
Packaging:
View all
Plastic Bottle
Catalog number 349030500
Price (USD)/ Each
83.00
-
Add to cart
Quantity:
50 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
83.00
Add to cart
1,2,4-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals
Catalog number349030500
Price (USD)/ Each
83.00
-
Add to cart
Chemical Identifiers
IUPAC Namebenzene-1,2,4-tricarboxylic acid
Molecular FormulaC9H6O6
InChI KeyARCGXLSVLAOJQL-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=O)C(=C1)C(O)=O
Molecular Weight (g/mol)210.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only