Thermo Scientific Chemicals

Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals

Catalog number: 349610250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals

Catalog number: 349610250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 349610250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-52-4
IUPAC Name
calcium dichloride
Molecular Formula
CaCl2
InChI Key
UXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ca++]
Appearance (Color)
White
Iron (Fe)
=<50 ppm
Barium (Ba)
=<200 ppm
Sulfate (SO4)
=<500 ppm
Magnesium (Mg)
=<1000 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text