Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals
Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 349615000
Price (USD)/ Each
64.65
Online exclusive
75.40 
Save 10.75 (14%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
75.40 
Save 10.75 (14%)
Add to cart
Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals
Catalog number349615000
Price (USD)/ Each
64.65
Online exclusive
75.40 
Save 10.75 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namecalcium dichloride
Molecular FormulaCaCl2
InChI KeyUXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ca++]
Molecular Weight (g/mol)110.98
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<50 ppm
Barium (Ba)=<200 ppm
Sulfate (SO4)=<500 ppm
Magnesium (Mg)=<1000 ppm
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only